Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam AICAR, AMPK Activator, Solid, >98% Purity, 50 mM/mL

Catalog No. 502704512
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mM/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4512 50 mM/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4512 Supplier Abcam Supplier No. AB14671350MM_ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable activator of AMP-activated protein kinase that is active in vivo and in vitro.

  • Molecular Weight: 258.23 Da
  • Purity: >98%
  • Form: Solid
  • Activity: Cell-permeable activator of AMP-activated protein kinase; active in vivo and in vitro
  • Mechanism: Taken up by adenosine transporters and phosphorylated to the active nucleotide ZMP, which mimics AMP effects on the AMPK system
  • Composition: 100% 5-Amino-1-β-D-ribofuranosyl-1H-imidazole-4-carboxamide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Shipping Condition Ambient
Target Molecule AMP-activated protein kinase
Buffer 100% 5-Amino-1-β-D-ribofuranosyl-1H-imidazole-4-carboxamide
Content And Storage Store at -20°C.
Form Solid
Formulation 100% 5-Amino-1-β-D-ribofuranosyl-1H-imidazole-4-carboxamide
Quantity 50 mM/mL
For Use With (Application) AMP-activated protein kinase activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.