Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam AICAR, Protein Kinase Activator, Liquid, >98% Purity, 50 mg

Catalog No. 502704561
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4561 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4561 Supplier Abcam Supplier No. AB14659450MGLQD
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable activator of AMP-activated protein kinase that mimics the effects of AMP on the AMPK system.

  • Molecular weight: 258.23 Da
  • Purity: >98%
  • Form: Liquid
  • Cell-permeable activator of AMP-activated protein kinase
  • Taken up into cells by adenosine transporters and phosphorylated to the active nucleotide ZMP
  • Mimics effects of AMP on the AMPK system
  • Active in vivo and in vitro
  • Storage buffer: 93.54% Water, 6.46% 5-Amino-1-β-D-ribofuranosyl-1H-imidazole-4-carboxamide
  • Storage temperature: -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule AMP-activated protein kinase
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Buffer 93.54% water, 6.46% 5-Amino-1-β-D-ribofuranosyl-1H-imidazole-4-carboxamide
Content And Storage Store at -20°C.
Form Liquid
Formulation 93.54% Water, 6.46% 5-Amino-1-β-D-ribofuranosyl-1H-imidazole-4-carboxamide
Quantity 50 mg
For Use With (Application) AMP-activated protein kinase activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.