Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIDA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7210394 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4111 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4111 Supplier Proteintech Group Inc Supplier No. 237241AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AIDA acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.

Specifications

Antigen AIDA
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AIDA
Gene Accession No. Q8C4Q6, Q96BJ3
Gene Alias AIDA, C1orf80, RP11 378J18.7
Gene Symbols AIDA
Host Species Rabbit
Immunogen AIDA Fusion Protein Ag20588
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108909, 64853
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.