Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534354
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534354 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534354 Supplier Invitrogen™ Supplier No. PA534354
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-34354 can be used with blocking peptide PEP-1397.

Androgen-induced 1 (AIG1) is a novel Pirh2-interacting protein, which was initially cloned from human dermal papilla cells and is evolutionally conserved from drosophila through to humans. It is a membrane protein and belongs to the protein family containing FAR-17a domain. AIG1 may play a role in androgen-regulated growth of hair follicles. AIG1 could serve as a new biomarker for the diagnosis and prognostic evaluation of HCCs and may be a novel candidate tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIG1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene AIG1
Gene Accession No. Q9D8B1, Q9NVV5
Gene Alias 1500031O19Rik; AIG1; AIG-1; androgen induced 1; androgen-induced 1; androgen-induced gene 1 protein; AV064870; AW413422; CGI-103; dJ95L4.1; FAHFA hydrolase AIG1; Fatty acid esters of hydroxy fatty acids hydrolase AIG1; RGD1562920
Gene Symbols AIG1
Host Species Rabbit
Immunogen A 17 amino acid synthetic peptide near the amino terminus of human AIG1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292486, 51390, 66253
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.