Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIM2 Polyclonal Antibody, eBioscience™
GREENER_CHOICE

Catalog No. 5013447
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-134-47 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-134-47 Supplier Invitrogen™ Supplier No. 14600893
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Description: This polyclonal antibody reacts with human, mouse, and rat AIM2 (also known as Antigen isolated from Immunoselected Melanoma 2). A member of the HIN-2000 family, this 40-kDa cytoplasmic protein is expressed in the spleen, small intestine, peripheral blood lymphocytes, and testis. Moreover, AIM2 is expressed in tumors of the neuroectodermis, breast, ovary, and colon. As a component of the inflammosome, AIM2 detects double-stranded viral and bacterial DNA present in the cytosol and initiates Caspase1 activation. Expression of AIM2 can be induced by interferons. Applications Reported: This polyclonal antibody has been reported for use in immunoblotting (WB). Applications Tested: This polyclonal antibody has been tested by immunoblotting of Jurkat cells and mouse macrophages treated for 12 hrs with IFN gamma and LPS. This can be used at less than or equal to a 1:1000 dilution. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

AIM2 is a member of the IFI20X /IFI16 family. It plays a putative role in tumorigenic reversion and may control cell proliferation. Interferon-gamma induces expression of AIM2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIM2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 1mg/mL BSA and 0.05% sodium azide
Gene AIM2
Gene Accession No. O14862, Q91VJ1
Gene Alias Absent in melanoma 2; AIM2; Gm1313; Ifi210; ifi-210; Interferon-inducible protein 210; interferon-inducible protein AIM2; Interferon-inducible protein p210; PYHIN4; RP11-520H16.3
Gene Symbols AIM2
Host Species Rabbit
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304987, 383619, 9447
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.