Learn More
AIMS™ CO2 Chamber
Catalog No. 14370180
Change view
Click to view available options
Type:
CO2 Chamber
Liner Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-370-180 CO2 Chamber
14-370-181 Liner Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-370-180 Supplier AIMS™ Supplier No. COIC
Add to Cart
Add to Cart
For mice and rats
Simple, quick and easy to use
  • Easy to see animals in chamber with see-through acrylic lid with door and quick connect with valve, 2.13m (7 ft.) tubing
  • 50 disposable chamber-liner bags collect animal feces and urine, keeping the chamber clean
  • 22.86cm (9 in.) diameter, easy to keep on bench
  • Allows user to place animals in chamber without losing CO2
  • Delivery tube within chamber fills chamber from bottom, decrease induction time – more humane; provides for ultra-quick animal induction
  • Solid sides prevent viewing of animals during processes
Order Info
Replacement disposable chamber-line bags available separately
Specifications
Type CO2 Chamber
Disposable Single-use
Includes Acrylic lid with door, 7 ft. tubing, 50 disposable insert bags
Diameter (English) 9 in.
Diameter (Metric) 22.86 cm