Learn More
AIMS™ Traditional Triangle Shaped Plastic Disposable Restraints
Description
Includes
Instructions and twist ties
This rodent restraint system is available in a variety of sizes and colors. The cone-shaped bags provide easier animal capture and increased animal comfort.
- Precut sizes for quick use
- Easy-Flex-PETM provides effortless wrapping and added animal comfort
- Body-Contour Cone-Shape provides easier restraint, exceptional comfort, and reduced animal stress
- Black and rose color bags reduce natural light and aids in reducing animal agitation and stress
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
Specifications
Specifications
| Color | Rose |
| For Use With (Application) | Tattooing, Tail Vein Injections, Oral Dosing, Decapitation and General Restraint |
| Type | Restraint |
| Disposable | Single-use |
| Material | Plastic |
| Shape | Triangle |
| Size | Small |
| Weight Range | ≤120 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.