Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ Traditional Triangle Shaped Plastic Disposable Restraints

Catalog No. 14370105
Change view
Click to view available options
Color:
Black
Clear
Rose
Size:
X-Small
Small
Medium
Large
Weight Range:
≤120 g
≤225 g
≤450 g
<20 g
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Weight Range
14-370-105 Rose Medium ≤225 g
14-370-100 Clear X-Small <20 g
14-370-102 Clear Medium ≤225 g
14-370-103 Clear Large ≤450 g
14-370-104 Rose Small ≤120 g
14-370-107 Black Small ≤120 g
14-370-108 Black Medium ≤225 g
14-370-101 Clear Small ≤120 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14-370-105 Supplier AIMS™ Supplier No. 1802R
Add to Cart
Add to Cart

Includes

Instructions and twist ties

Humane, safe and disposable rodent restraint system

This rodent restraint system is available in a variety of sizes and colors. The cone-shaped bags provide easier animal capture and increased animal comfort.

  • Precut sizes for quick use
  • Easy-Flex-PETM provides effortless wrapping and added animal comfort
  • Body-Contour Cone-Shape provides easier restraint, exceptional comfort, and reduced animal stress
  • Black and rose color bags reduce natural light and aids in reducing animal agitation and stress

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.

Specifications

Color Rose
For Use With (Application) Tattooing, Tail Vein Injections, Oral Dosing, Decapitation and General Restraint
Type Restraint
Disposable Single-use
Material Plastic
Shape Triangle
Size Medium
Weight Range ≤225 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.