Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimScreen™ MultiDrug Urine DipDevice

Catalog No. 23111327
Change view
Click to view available options
Format:
10 Drug Screen
12 Drug Screen
2 Drug Screen
3 Drug Screen
4 Drug Screen
5 Drug Screen
Detectable Analytes:
Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Cannibinoids (THC), Cocaine (COC), Methylenedioxymethamphetamine (MDMA), Methodone (MTD), Methamphetamine (mAMP), Opiates (OPI), Oxycodone (OXY), Phencyclidine (PCP), Propoxyphene (PPX)
Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Cannibinoids (THC), Cocaine (COC), Methylenedioxymethamphetamine (MDMA), Methodone (MTD), Methamphetamine (mAMP), Opiates (OPI), Phencyclidine (PCP)
Amphetamine (AMP), Cocaine (COC), Cannibinoids (THC), Methamphetamine (mAMP), Opiates (OPI)
Amphetamine (AMP), Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Phencyclidine (PCP)
Cannibinoids (THC), Cocaine (COC)
Cannibinoids (THC), Cocaine (COC), Methamphetamine (mAMP)
Cannibinoids (THC), Cocaine (COC), Methamphetamine (mAMP), Opiates (OPI)
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Detectable Analytes
23-111-327 5 Drug Screen Amphetamine (AMP), Cocaine (COC), Cannibinoids (THC), Methamphetamine (mAMP), Opiates (OPI)
23-111-323 2 Drug Screen Cannibinoids (THC), Cocaine (COC)
23-111-324 3 Drug Screen Cannibinoids (THC), Cocaine (COC), Methamphetamine (mAMP)
23-111-325 4 Drug Screen Cannibinoids (THC), Cocaine (COC), Methamphetamine (mAMP), Opiates (OPI)
23-111-326 5 Drug Screen Amphetamine (AMP), Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Phencyclidine (PCP)
23-111-328 10 Drug Screen Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Cannibinoids (THC), Cocaine (COC), Methylenedioxymethamphetamine (MDMA), Methodone (MTD), Methamphetamine (mAMP), Opiates (OPI), Phencyclidine (PCP)
23-111-329 12 Drug Screen Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Cannibinoids (THC), Cocaine (COC), Methylenedioxymethamphetamine (MDMA), Methodone (MTD), Methamphetamine (mAMP), Opiates (OPI), Oxycodone (OXY), Phencyclidine (PCP), Propoxyphene (PPX)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 23-111-327 Supplier Germaine™ Laboratories Supplier No. 45625
Add to Cart
Add to Cart

Germaine™ Laboratories AimScreen™ MultiDrug Urine DipDevice is competitive binding, lateral flow immunochromatographic assays for qualitative and simultaneous detection of drugs in human urine

  • Rapidly screen for broad range of illicit and prescription drugs including:
    • Amphetamine (AMP)
    • Barbituates (BAR)
    • Benzodiazepine (BZO)
    • Cannabinoids (THC)
    • Cocaine (COC)
    • Ecstasy (MDMA)
    • Methadone (MDT)
    • Methamphetamine (MET)
    • Opiates/Morphine (OPI)
    • Oxycodone (OXY)
    • Phenocyclidine (PCP)
    • Propoxyphene (PPX)
  • 510(k) Cleared and CLIA-waived
  • Simple procedure: collect, dip and read-results in as little as five minutes
  • Test device remains in control of administrator

Specifications

Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Amphetamine (AMP), Cocaine (COC), Cannibinoids (THC), Methamphetamine (mAMP), Opiates (OPI)
Format 5 Drug Screen
Sample Type Urine
Type Urine DipDevice
Disposable Single-use
Quantity 25 Tests
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.