Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimScreen™ Urine DipDevice

Catalog No. 23111343
Change view
Click to view available options
Detectable Analytes:
Amphetamine (AMP)
Barbiturates (BAR)
Benzodiazepines (BZO)
Buprenorphine (BUP)
Cannibinoids (THC)
Cocaine (COC)
Methamphetamine (mAMP)
Methodone (MTD)
Opiates (OPI)
Opiates (OPI), Morphine (MOP)
Oxycodone (OXY)
Phencyclidine (PCP)
Tricyclic Antidepressants (TCA)
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-111-343 Tricyclic Antidepressants (TCA)
23-111-338 Opiates (OPI)
23-111-341 Phencyclidine (PCP)
23-111-339 Opiates (OPI), Morphine (MOP)
23-111-340 Oxycodone (OXY)
23-111-333 Buprenorphine (BUP)
23-111-335 Cocaine (COC)
23-111-331 Methamphetamine (mAMP)
23-111-332 Barbiturates (BAR)
23-111-334 Benzodiazepines (BZO)
23-111-337 Methodone (MTD)
23-111-344 Cannibinoids (THC)
23-111-330 Amphetamine (AMP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 23-111-343 Supplier Germaine™ Laboratories Supplier No. 44225
Add to Cart
Add to Cart

Germaine™ Laboratories AimScreen™ Urine DipDevice is competitive binding, lateral flow immunochromatographic assays for qualitative and simultaneous detection of various drugs in human urine

  • Rapidly screen for broad range of illicit and prescription drugs
  • 510(k) Cleared and CLIA-waived
  • Simple procedure: collect, dip and read-results in as little as five minutes
  • Test device remains in control of administrator

Specifications

Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Tricyclic Antidepressants (TCA)
Sample Type Urine
Type Urine DipDevice
Disposable Single-use
Quantity 25 Tests
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.