Learn More
Germaine™ Laboratories AimStrip™ Tandem Triglycerides Control Solution
Catalog No. 23111386
Change view
Click to view available options
Quantity:
2 x 3 mL
Catalog No. Quantity
23-111-386 2 x 3 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-111-386 Supplier Germaine™ Laboratories Supplier No. 77412
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used to test the precision of the AimStrip™ Tandem Lipid Profile Measuring System and to detect systematic analytic deviations that may arise from reagent or analytical variation.
  • Control Solution for AimStrip™ Tandem Triglycerides Test Strips
  • Simply insert test, apply specimen & read results
Specifications
Detectable Analytes Triglycerides
For Use With (Equipment) AimStrip™ TANDEM Measuring System
Type Quality Control Solution
Includes Contol Solution Level 1 (3 mL), Contol Solution Level 2 (3 mL), 10 Capillary Rods
Level Bi-level
Quantity 2 x 3 mL