Learn More
MP Biomedicals™ AIN-76A Semipurified Diet
AIN-76A Semipurified Diet is intended for growth and maintenance of animals during the first year of life.
Supplier:  MP Biomedicals™ 0296009720
 View more versions of this product
Catalog No. 02-960-09720
Only null left
Add to Cart
Description
Description
  • Contains casein purified high nitrogen, DL-methionine, sucrose, corn starch, Alphacel (non-nutritive bulk), corn oil, choline bitartrate, AIN mineral mix, and a 10-fold increase in the amount of vitamin K source to 500 μg/kg.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pellets
20 kg
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.