Learn More
Bio-Serv™ AIN-93G Rodent Diet

Purified diet formulated to meet the nutritional requirements of growing and gestating rodents
Supplier: Bio-Serv™ F3156

Description
- Widely used and published
- Phytoestrogen free
- Chlorophyll free—does not interfere with imaging studies
- Nutritionally assayed
- Ingredients: Corn Starch, Casein, Maltodextrin, Sucrose, Soybean Oil, Cellulose, Mineral Mix, Vitamin Mix, L-Cystine, Choline Bitartrate, tBHQ
Specifications
Rodent Diet | |
Pellet | |
Pellet | |
Rodent | |
1.27 cm |
Corn Starch, Casein, Maltodextrin, Sucrose, Soybean Oil, Cellulose, Mineral Mix, Vitamin Mix, L-Cystine, Choline Bitartrate, tBHQ | |
Used and published for research objectives | |
10 x 1 kg | |
0.5 in. |
Safety and Handling
ShelfLife : 6 Months
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.