Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIP Monoclonal Antibody (10G8)
GREENER_CHOICE

Catalog No. PIMA549238
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549238 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549238 Supplier Invitrogen™ Supplier No. MA549238
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human ARA9 shares 95% amino acid (aa) sequence identity with both mouse and rat ARA9. Positive Control - WB: human HELA whole cell, human HEPG2 whole cell, human placenta tissue, human CACO-2 whole cell, human U20S whole cell, monkey COS-7 whole cell, human K562 whole cell, rat heart tissue, rat skeletal muscle tissue, rat brain tissue, rat spleen tissue, mouse brain tissue, mouse HEPA1-6 whole cell. IHC: human intestinal cancer tissue, human tonsil tissue, rat spleen tissue. ICC/IF: A431 cell. Flow: A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIP
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10G8
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene Aip
Gene Accession No. O00170, O08915, Q5FWY5
Gene Alias AA408703; AH receptor-interacting protein; AIP; ARA9; aryl hydrocarbon receptor interacting protein; aryl-hydrocarbon receptor-interacting protein; AW476050; D19Bwg1412e; FKBP16; FKBP37; HBV X-associated protein 2; HBV-X associated protein; immunophilin homolog ARA9; Immunophilin XAP2; SMTPHN; XAP2; XAP-2
Gene Symbols Aip
Host Species Mouse
Immunogen E.coli-derived human ARA9 recombinant protein (Position: D91-H330).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11632, 282827, 9049
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.