Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578355 Supplier Invitrogen™ Supplier No. PA578355
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: human WWP2-transfected 293T Predicted Reactivity: Mouse (90%), Rat (90%), Dog (89%), Rhesus Monkey (96%), Chimpanzee (99%), Bovine (87%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the NEDD4-like protein family. The family of proteins is known to possess ubiquitin-protein ligase activity. The encoded protein contains 4 tandem WW domains. The WW domain is a protein motif consisting of 35 to 40 amino acids and is characterized by 4 conserved aromatic residues. The WW domain may mediate specific protein-protein interactions. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene WWP2
Gene Accession No. O00308
Gene Alias 1300010O06Rik; AA690238; AIP2; atrophin-1 interacting protein 2; Atrophin-1-interacting protein 2; AW554328; HECT-type E3 ubiquitin transferase WWP2; id:ibd1121; NEDD4-like E3 ubiquitin-protein ligase WWP2; wu:fo87e10; WW domain containing E3 ubiquitin protein ligase 2; WW domain-containing protein 2; WW domain-containing protein 4; WWP2; WWp2-like; zgc:154036
Gene Symbols WWP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AIP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11060
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.