Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PureAire Monitoring Systems PureAire Monitoring Systems Oxygen Monitor with 10 ft. Remote Sensor
SDP

Catalog No. 18222007
Change view
Click to view available options
Enclosure Material:
Polycarbonate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Enclosure Material
18-222-007 Polycarbonate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-222-007 Supplier PureAire Monitoring Systems Supplier No. 99028
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PureAire Monitoring Systems Air Check Oxygen Monitor with remote sensing capabilities

  • Perfect for use in a vacuum environment, OLED printer, nitrogen generator, or machines using nitrogen, and cryogenics
  • Sensor is connected to the monitor using a 3 meter cable for easy connection.
  • Remote controller can operate monitor at distances of up to 1,000 meters (0.6 miles)
  • Long-life zirconium sensor with average life of 10+ years
  • Two-Alarm setpoints for 19.5% and 18.0%
  • Built-in audible alarm 90 db
  • Digital display, and 4-20mA analog output
  • Wall mounting brackets
  • Remote Sensor up 10 ft away
  • CE marked. UL, and CUL approval
  • Simply secure the unit on the wall 1-2 meters from the gas source, plug it in, and power it on.
  • Levels are displayed clearly on the screen for easy access and visibility

Specifications

Temperature (Metric) Operating -40°C to +55°C
Accuracy ±1% of Full Scale
Calibration Factory Calibrated against a NIST Traceable Reference Standard
Certifications/Compliance CE Approval
Depth (English) 3.3 in.
Depth (Metric) 82.5 mm
Electrical Requirements 24 VDC
Enclosure Material Polycarbonate
Height (English) 4.5 in.
Height (Metric) 114.3 mm
Outputs 4-20 mA
Weight (English) 1.6 lb.
Weight (Metric) 0.8 kg
Width (English) 6.5 in.
Width (Metric) 165.1 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.