Learn More
Grainger AIR DUCT 12IN DIA X 25FT
Supplier:  Grainger 39FW30
Encompass_Preferred
The Air Systems International 12" Diameter Air Duct is a galvanized steel circular duct ideal for HVAC and exhaust ventilation applications. It features a 26 gauge thickness with a smooth interior for efficient airflow. The ducts have interlocking connections for easy assembly and can be cut to desired lengths. Diameters range from 4" to 16". These air ducts meet SMACNA standards and offer corrosion resistance. With various diameters and connection options, these galvanized steel ducts are versatile for commercial and residential HVAC installations.
Catalog No. NC2602732
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.