Learn More
Thermo Scientific™ Air Filter Kits
GSA_VA
Catalog No. 13265247
Change view
Click to view available options
For Use With (Equipment):
Polar Accel 250 and 500
ThermoFlex 2500
ThermoFlex 3500 or 5000
ThermoFlex 7500 or 1000
ThermoFlex 900 or 1400
Type:
Air Filter
Air and Fluid Filter
Size:
100 micron
35 micron
N/A
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type Size
13-265-247 ThermoFlex 900 or 1400 Air and Fluid Filter 35 micron
13-265-248 ThermoFlex 2500 Air and Fluid Filter 100 micron
13-265-249 ThermoFlex 2500 Air and Fluid Filter 35 micron
13-265-250 ThermoFlex 3500 or 5000 Air and Fluid Filter 100 micron
13-265-251 ThermoFlex 3500 or 5000 Air and Fluid Filter 35 micron
13-100-768 ThermoFlex 7500 or 1000 Air and Fluid Filter 100 micron
13-265-252 ThermoFlex 7500 or 1000 Air and Fluid Filter 35 micron
13-100-782 Polar Accel 250 and 500 Air Filter N/A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 13-265-247 Supplier Thermo Scientific™ Supplier No. 598000000005
Add to Cart
Add to Cart
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific recirculating chillers.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ThermoFlex 900 or 1400
Type Air and Fluid Filter
Size 35 micron