Learn More
Cytiva 37 mm Air Monitoring Cassettes - Three-piece unit, unassembled (100/pkg)
Supplier:  Cytiva 4339N
Encompass
Ideal for general particulate air sampling when fitted with the appropriate membrane or filter.•Designed with NIOSH requirements in mind•Ideal for air sampling•Styrene acrylonitrile housingStyrene acrylonitrile housing consists of inlet, outlet, and inner ring for open- or closed-face monitoring. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3038