Learn More
Scott Safety™ Air-Pak™ X3 SCBA
Description
Designed with enhancements to comfort, hose and wire management, weight distribution, fire ground communications and serviceability.
- Fire ground communications enhanced with the introduction of the AV-3000 HT facepiece, EPIC 3 Communication and optimally positioned “buddy” lights
- “External” HUD for ease in air status updates of the team
- Dual redundant pressure reducer
- Vibralert tactile alarm
- Pak Alert PASS (Personal Alert Safety System) - standardized sound signals man-down situation
- Pak Tracker Firefighter Locator Systems with HHR (handheld receiver) - alerts remote users of man-down situation; directional HHR provides direction for rescue teams
- SEMS II - includes Pak Alert PASS and PAK Alert locator features plus redundant network to carry PASS and low-air notifications, reducing radio traffic
Specifications
Specifications
| Certifications/Compliance | 1981, 2007, 2013 NFPA |
| Product Type | SCBA |
| Pressure | 5500 psi |
| For Use With (Application) | Fire ground communications are enhanced with the introduction of the AV-3000™ HT facepiece, Epic 3 communication and optimally positioned “buddy” lights “external” hud |
| Harness Style | Standard |
| Hose Type | Quick-Connect |
| Includes | Duall EBSS; Snap-Change cylinder connection; Pak-Tracker Locator |