Learn More
Corning™ PYREX™ Air Reflux Condenser With 14/10 Threaded Joint
Catalog No. 11850107 Shop All Corning Products
Change view
Click to view available options
Standard Taper Joint No:
14/10 Bottom, 7/10 Top joint
Catalog No. Standard Taper Joint No
11-850-107 14/10 Bottom, 7/10 Top joint
1 options
Catalog No. 11-850-107 Supplier Corning™ Supplier No. 285014
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
For use with Pyrex microchemistry kits
Replacement part for 6949M series of PYREX microchemistry kits.
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Quantity 6/Cs.
For Use With (Equipment) Pyrex microchemistry kits
Standard Taper Joint No 14/10 Bottom, 7/10 Top joint
Type Air Reflux Condenser