Learn More
AirClean™ Systems Activated Bonded Carbon Chemisorptive Filters
SDP
Catalog No. 361018825 Shop All AirClean Systems Products
Change view
Click to view available options
Filter Type:
ACF100
ACF100H
ACF300
ACF400
ACFFOR
ACFGLT
For Use With (Application):
Ammonia or Amines
Formaldehyde
Glutaraldehyde
Inorganic Acids
Organic Solvent Vapors
Organic Vapor, Solvents, and Odors
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type For Use With (Application)
36-101-8825 ACF100 Organic Vapor, Solvents, and Odors
36-101-8827 ACF300 Ammonia or Amines
36-101-8828 ACF400 Inorganic Acids
36-101-8829 ACFFOR Formaldehyde
36-101-8843 ACF100H Organic Solvent Vapors
36-101-8830 ACFGLT Glutaraldehyde
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 36-101-8825 Supplier AirClean™ Systems Supplier No. ACF100
Add to Cart
Add to Cart
For the removal of fumes and vapors
  • Coconut shell activated carbon
  • Chemically impregnated carbon for increased load capacity
  • Large surface area and highly developed internal structure allows adsorption up to 50%
  • Bonded design eliminates carbon dusting and prevents carbon shift that can lead to “dead spots”
Specifications
Filter Type ACF100
Material Activated Carbon
For Use With (Application) Organic Vapor, Solvents, and Odors