Learn More
AirClean™ Systems Bonded Carbon Filters
SDP
Catalog No. 361018833 Shop All AirClean Systems Products
Change view
Click to view available options
For Use With (Application):
For Ammonia or Amines
For Inorganics
For organic solvent vapors
Filter Type:
ACF100-18
ACF209-18
ACF300-18
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Filter Type
36-101-8833 For organic solvent vapors ACF100-18
36-101-8835 For Ammonia or Amines ACF300-18
36-101-8834 For Inorganics ACF209-18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 36-101-8833 Supplier AirClean™ Systems Supplier No. ACF10018
Add to Cart
Add to Cart
AirClean™ Systems Bonded Carbon Filters used in evidence drying cabinets to absorb the fumes and odors.
  • Used in evidence drying cabinets to absorb fumes and odors associated with drying and decay of bloody articles
Specifications
Filter Type ACF100-18
For Use With (Equipment) AC648 Units with Negative Pressure
Weight (English) 15 lb.
Weight (Metric) 6.8 Kg
Dimensions 30.4 x 45.7 x 7.62 cm (12 x 18 x 3 in.)
Product Type Filter
For Use With (Application) For organic solvent vapors