Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AirClean™ Systems Fume Hood Fixtures
SDP

Catalog No. 361000147 Shop All AirClean Systems Products
Change view
Click to view available options
Color:
Brown
Green
Orange
For Use With (Application):
For Air
For Cold Water
For Nitrogen
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Application)
36-100-0147 Orange For Air
36-100-0151 Green For Cold Water
36-100-0154 Brown For Nitrogen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 36-100-0147 Supplier AirClean™ Systems Supplier No. ACAFXAR
Add to Cart
Add to Cart

Available on all sliding sash ductless fume hoods and AirMax™ polypropylene total exhaust fume hoods. AirClean™ Systems Fume Hood Fixtures are installed and color coordinated, per international standards.

  • Available on all sliding sash ductless fume hoods and AirMax™ polypropylene total exhaust fume hoods
  • Installed and color coordinated, per international standards, on each post
  • All fixtures are pre-plumbed and wired for easy laboratory installation

Specifications

For Use With (Equipment) All sliding sash ductless fume hoods and AirMax polypropylene total exhaust fume hoods
Product Type Fixture
For Use With (Application) For Air
Color Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.