Learn More
AirClean™ Systems HEPA Filters
SDP
Catalog No. 361018805 Shop All AirClean Systems Products
Change view
Click to view available options
For Use With (Equipment):
AC5000HLF and UV
DS200, AC648H, AC6400A
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
36-101-8805 DS200, AC648H, AC6400A
36-101-8811 AC5000HLF and UV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-101-8805 Supplier AirClean™ Systems Supplier No. ACFHEPA18
Add to Cart
Add to Cart
High efficiency particulate air (HEPA) filter, AirClean™ systems HEPA filters can be used with ductless fume hoods and drying cabinets.
  • High efficiency particulate air (HEPA) filters with 99.997% efficiency rating for collection at 0.3 microns
  • Used for containment of particles - toxic powders, asbestos, lead, fingerprint powders
Specifications
Filter Type HEPA
For Use With (Equipment) DS200, AC648H, AC6400A
Product Type Filter