Learn More
AirClean™ Systems Polypropylene Total Exhaust Fume Hoods
To maximize workspace, choose the AirClean Systems Polypropylene Total Exhaust Fume Hood. It's single wall design allows for more room to do actual work.
Supplier: AirClean™ Systems AC4000TE
Description
- Comes with folding sash and vapor-proof fluorescent light
- AirZone™ baffling allows for predictable and effective airflow pattern to capture toxic chemical fumes and vapors
- Thermally fused propylene design provides built-in work base that will effectively contain liquids and provide excellent chemical resistance
- Seamless construction allows for easy cleaning
- Optional airflow monitor for constant alphanumeric readout of airflow
- Available with single or dual exhaust duct connections
- Polypropylene base cabinets available in vented and non-vented models
Specifications
| Fume Hood | |
| Polypropylene | |
| 36 in. | |
| 32 x 47 x 34 in. (81.2 x 119.3 x 86.3 cm) | |
| 149.8 cm | |
| 48 in. |
| Vapor-proof Fluorescent | |
| Total exhaust, Single wall for more work space, Corrosion-resistant | |
| 91.4 cm | |
| 59 in. | |
| Folding sash; AirZone baffling; Single/dual exhaust duct connections; Vapor-proof fluorescent light | |
| 121.9 cm |