Learn More
Electron Microscopy Sciences Air-Evac Bottle, 1qt Size
SDP
Supplier:  Electron Microscopy Sciences 74356
Encompass_Preferred
For storing chemicals that are air and light sensitive. Suitable for use with all photographic chemicals. As chemicals are used, Air-Evac can be compressed forcing out extraneous air from the bottle contents. Cap is then replaced for an air tight seal.
Catalog No. 50-244-74