Learn More
Med Vet International Berman Airway, White, 90 mm, 12/Box
Supplier:  Med Vet International DYND60420
Encompass_Preferred
Berman Airway: Slotted sides permit passage of suction catheters without obstructing the air passage. Triple-purpose airway serves as a bite block and tongue depressor to avoid crimping and protect teeth. Disposable, single-use Berman airway automatically curves to the contour of the pharynx. Smooth rounded tip reduces trauma to upper airway, and the flat surface helps ensure patient comfort. 90 mm, Medium Adult. 12/bx.
Catalog No. 50-283-2371
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.