Learn More
Med Vet International Dynarex Nasopharyngeal Airway Kits - 6 Npa + 6 Packs Of Jelly, Each
Supplier:  Med Vet International 36270
Encompass_Preferred
The Dynarex Nasopharyngeal Airway is a high-quality tubular device designed to help keep patients’ airways open for improved oxygenation. The Dynarex Nasopharyngeal Airway features a fixed flange end to limit the depth of insertion and internal striations to allow for smoother passage of suction catheters. Dynarex Nasopharyngeal Airway Kits include an assortment of airways in multiple sizes to accommodate patients’ specific needs, and packets of lubrication jelly for added convenience. Each kit comes with either 6 or 9 airways for ultimate versatility. The Dynarex Nasopharyngeal Airway is designed for nasal insertion to help create and maintain an open and secure airway. The Dynarex Nasopharyngeal Airway features a fixed flange end for safer insertion and internal striations for optimal suctioning efficiency. Dynarex Nasopharyngeal Airway Kits include an assortment of airways in multiple sizes for optimal versatility and packets of lubrication jelly for added convenience. 
Catalog No. 50-283-1443
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.