Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AK4 Monoclonal Antibody (OTI5B10), TrueMAB™, OriGene
SDP

Catalog No. p-200010161
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1237 30 μL
50-167-3898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1237 Supplier OriGene Supplier No. TA502899S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AK4 Monoclonal antibody specifically detects AK4 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen AK4
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI5B10
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FGF21
Gene Accession No. Q9NSA1
Gene Alias adenylate kinase 3 alpha-like 1; Adenylate kinase 3-like; adenylate kinase 3-like {ECO:0000255; adenylate kinase 3-like 1; adenylate kinase 3-like 2; adenylate kinase 4; adenylate kinase 4, mitochondrial; adenylate kinase 4, mitochondrial {ECO:0000255; Adenylate kinase isoenzyme 4; adenylate kinase isoenzyme 4, mitochondrial; adenylate kinase isoenzyme 4, mitochondrial; adenylate kinase 4, mitochondrial; AK 4; AK 4 {ECO:0000255; AK3; Ak-3; Ak3b; Ak3l1; AK3L2; AK4; Ak-4; ATP-AMP transphosphorylase; D4Ertd274e; GTP:AMP phosphotransferase AK4; GTP:AMP phosphotransferase AK4 {ECO:0000255; GTP:AMP phosphotransferase AK4, mitochondrial; HAMAP-Rule:MF_03170}; mitochondrial adenylate kinase-3; nucleoside-triphosphate-adenylate kinase; RP4-686B20.1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 29-209 of human FGF21 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26291
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.