Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984235
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4235 Supplier Bioss Supplier No. BS18605R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes an enzyme which reversibly catalyzes the phosphorolysis of purine nucleosides. The enzyme is trimeric, containing three identical subunits. Mutations which result in nucleoside phosphorylase deficiency result in defective T-cell (cell-mediated) immunity but can also affect B-cell immunity and antibody responses. Neurologic disorders may also be apparent in patients with immune defects. A known polymorphism at aa position 51 that does not affect enzyme activity has been described. A pseudogene has been identified on chromosome 2.

Specifications

Antigen AKAP1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AKAP1
Gene Accession No. Q92667
Gene Alias A kinase (PRKA) anchor protein 1; a kinase anchor protein 1, mitochondrial; Akap; AKAP 149; Akap1; AKAP121; AKAP149; AKAP84; A-kinase anchor protein 1, mitochondrial; A-kinase anchor protein 149 kDa; A-kinase anchoring protein 1; C76494; C81186; DAKAP1; D-AKAP1; D-AKAP-1; Dual specificity A-kinase-anchoring protein 1; dual-specificity A-kinase anchoring protein 1; PPP1R43; PRKA1; protein kinase A anchoring protein 1; protein kinase A1; Protein kinase A-anchoring protein 1; protein phosphatase 1, regulatory subunit 43; S-AKAP; SAKAP84; S-AKAP84; spermatid A-kinase; Spermatid A-kinase anchor protein; Spermatid A-kinase anchor protein 84; TDRD17; testicular secretory protein Li 5; tudor domain containing 17
Gene Symbols AKAP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AKAP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8165
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.