Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114833
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114833 Supplier Invitrogen™ Supplier No. PA5114833
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AKAP12.

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is expressed in endothelial cells, cultured fibroblasts, and osteosarcoma cells. It associates with protein kinases A and C and phosphatase, and serves as a scaffold protein in signal transduction. This protein and RII PKA colocalize at the cell periphery. This protein is a cell growth-related protein. Antibodies to this protein can be produced by patients with myasthenia gravis. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene AKAP12
Gene Accession No. Q02952
Gene Alias A kinase (PRKA) anchor protein (gravin) 12; A kinase (PRKA) anchor protein 12; AI317366; AKAP 250; AKAP12; AKAP-12; AKAP12A; AKAP12B; AKAP12G; AKAP250; A-kinase anchor protein 12; A-kinase anchor protein 250 kDa; A-kinase anchor protein, 250kDa; A-kinase anchoring protein 12; Gag12; gercelin; germ cell lineage protein gercelin; Gravin; kinase scaffold protein gravin; Myasthenia gravis autoantigen; myasthenia gravis autoantigen gravin; Srcs5; Src-Suppressed C Kinase Substrate; SSeCKS; SSeCKS, Gravin, AKAP250; testis specific gene A12; Tsga12; v-src suppressed transcript 5
Gene Symbols AKAP12
Host Species Rabbit
Immunogen A synthesized peptide derived from human AKAP12(Accession Q02952), corresponding to amino acid residues G1749-S1782.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9590
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.