Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP13/AKAP-Lbc Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140851
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0299 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0299 Supplier Bethyl Laboratories, Inc Supplier No. A301404AT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The type II cAMP-protein kinase (PKA) is a multifunctional kinase with a broad range of substrates. Specificity of PKA signaling is thought to be mediated by the compartmentalization of the kinase to specific sites within the cell. To maintain this specific localization, the R subunit (RII) of PKA interacts with specific RII-anchoring proteins. The family of RII-anchoring proteins has been designated A-kinase anchoring proteins (AKAP). AKAP 13, also known as Brx (breast cancer nuclear receptor-binding auxiliary protein), Lbc (lymphoid blast crisis oncogene), HA-3 or Ht31 (human thyroid-anchoring protein 31), functions as a cAMP-dependent scaffold anchor for PKA and also has Rho-GEF activity. It is known to regulate TLR2 signaling, NFkB activation, protein kinase D activation and participate in Actin stress fiber formation. Seven isoforms exist for AKAP 13 and, depending on the isoform, it localizes to the cytoplasm, nucleus or cell membrane.

Specifications

Antigen AKAP13/AKAP-Lbc
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene AKAP13
Gene Accession No. Q12802
Gene Alias A-kinase anchor protein 13, AKAP-13, AKAP-Lbc, Breast cancer nuclear receptor-binding auxiliary protein, Guanine nucleotide exchange factor Lbc, Human thyroid-anchoring protein 31, Lymphoid blast crisis oncogene, LBC oncogene, Non-oncogenic Rho GTPase-specific GTP exchange factor, Protein kinase A-anchoring protein 13, PRKA13, p47, BRX, HT31, LBC, AKAP13
Gene Symbols AKAP13
Host Species Rabbit
Immunogen Between 2754 and 2804
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11214
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.