Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104533
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5104533 100 μL
1 options

Catalog No. PIPA5104533

Supplier: Invitrogen™ PA5104533

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AKAP2.

Binds to regulatory subunit (RII) of protein kinase A. May be involved in establishing polarity in signaling systems or in integrating PKA-RII isoforms with downstream effectors to capture, amplify and focus diffuse, trans-cellular signals carried by cAMP. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AKAP2
Gene Accession No. Q9Y2D5
Gene Alias A kinase (PRKA) anchor protein 2; AA959716; AI649048; AKAP expressed in kidney and lung; AKAP2; AKAP-2; AKAPKL; AKAP-KL; A-kinase anchor protein 2; A-kinase anchoring protein 2; B230340M18Rik; KL2B; MISP2; PALM2-AKAP2; PRKA2; protein kinase A anchoring protein 2; protein kinase A2; protein kinase A-anchoring protein 2
Gene Symbols AKAP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human AKAP2(Accession Q9Y2D5), corresponding to amino acid residues S78-A128.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11217
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.