Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKAP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576600
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576600 Supplier Invitrogen™ Supplier No. PA576600
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes a member of the A-kinase anchoring protein family, a group of functionally related proteins that bind to a regulatory subunit of cAMP-dependent protein kinase A and target the enzyme to specific subcellular compartments. AKAPs have a common RII-binding domain, but contain different targeting motifs responsible for directing PKA to distinct intracellular locations. Three alternatively spliced transcript variants encoding different isoforms have been described. Additional variants exist, but their full-length natures have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene AKAP7
Gene Accession No. O43687, O55074, Q6JP77, Q9P0M2
Gene Alias 6430401D08; A kinase (PRKA) anchor protein 7; AI662165; AKAP 18; AKAP 9; AKAP15; AKAP18; AKAP-18; AKAP-18 {ECO:0000250; AKAP18d; Akap7; akap7 {ECO:0000312; AKAP-7 isoform alpha; AKAP-7 isoform gamma; AKAP-7 isoforms alpha and beta; AKAP-7 isoforms delta and gamma; A-kinase anchor protein 18; A-kinase anchor protein 18 {ECO:0000250; A-kinase anchor protein 18 kDa; A-kinase anchor protein 7; A-kinase anchor protein 7 isoform alpha; A-kinase anchor protein 7 isoform gamma; A-kinase anchor protein 7 isoforms alpha and beta; A-kinase anchor protein 7 isoforms delta and gamma; A-kinase anchor protein 9 kDa; A-kinase anchoring protein 18 ,isoform delta; A-kinase anchoring protein 7; BB170514; PRKA7 isoform alpha; PRKA7 isoform gamma; PRKA7 isoforms alpha/beta; PRKA7 isoforms delta and gamma; protein kinase A anchoring protein 7; protein kinase A-anchoring protein 7 isoform alpha; Protein kinase A-anchoring protein 7 isoform gamma; Protein kinase A-anchoring protein 7 isoforms alpha/beta; Protein kinase A-anchoring protein 7 isoforms delta and gamma; RGD:1303071}; UniProtKB:F8VQ58}
Gene Symbols AKAP7
Host Species Rabbit
Immunogen Recombinant fusion protein of human AKAP7.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361458, 432442, 9465
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.