Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP9/CG-NAP Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560937
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0937 Supplier Bethyl Laboratories, Inc Supplier No. A301662A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The A-kinase anchor proteins are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternate splicing of this gene results in at least two isoforms that localize to the centrosome and the Golgi apparatus, and interact with numerous signaling proteins from multiple signal transduction pathways. These signaling proteins include type II protein kinase A, serine/threonine kinase protein kinase N, protein phosphatase 1, protein phosphatase 2a, protein kinase C-epsilon and phosphodiesterase 4D3.

Specifications

Antigen AKAP9/CG-NAP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene AKAP9
Gene Accession No. Q99996
Gene Alias AKAP9, CG-NAP, PRKA9, YOTIAO, AKAP350, HYPERION, KIAA0803, A kinase (PRKA) anchor protein (yotiao) 9, AKAP120-like protein, kinase N-associated protein, A-kinase anchoring protein 450, A-kinase anchor protein, 350kDa, protein kinase A anchoring protein 9, centrosome- and golgi-localized protein, AKAP450, MU-RMS-40.16A, A-kinase anchor protein 9, AKAP9-BRAF fusion protein
Gene Symbols AKAP9
Host Species Rabbit
Immunogen Between 1075 and 1125
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10142
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.