Learn More
TARGETMOL CHEMICALS INC AKB-6899 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T29797
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AKB-6899 is an inhibitor of prolyl hydroxylase domain 3 (PHD3) and increases the soluble form of the VEGF receptor (sVEGFR-1) production from GM-CSF-treated macrophages. AKB-6899 leads to stabilization of HIF-2(alpha) which induces sVEGFR-1 production from tumor-associated macrophages and decreases tumor growth. purity: 98%
Catalog No. 50-258-0447
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.