Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKR1C1/AKR1C2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578752
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578752 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578752 Supplier Invitrogen™ Supplier No. PA578752
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human A549 whole cell, human HCCP tissue, rat liver tissue, rat kidney tissue, mosue liver tissue, mosue kidney tissue. ICC/IF: HepG2 cell. Flow: HepG2 cell.

This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols using NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme binds bile acid with high affinity, and shows minimal 3-alpha-hydroxysteroid dehydrogenase activity. And this gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR1C1/AKR1C2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene AKR1C1
Gene Accession No. P52895, Q04828, Q6AYQ2
Gene Alias 17-alpha-HSD; 17-alpha-hydroxysteroid dehydrogenase; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase); 20-ALPHA-HSD; 20-alpha-hydroxysteroid dehydrogenase; 2-ALPHA-HSD; 3(or 17)-alpha-hydroxysteroid dehydrogenase; 3-alpha-HSD3; 3-alpha-hydroxysteroid dehydrogenase; AKR1C1; Akr1c2; Akr1c21; Akr1c6; AKR1C-pseudo; aldo-keto reductase C; aldo-keto reductase family 1 member C1; Aldo-keto reductase family 1 member C2; aldo-keto reductase family 1 member C21; aldo-keto reductase family 1, member C1; aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase); aldo-keto reductase family 1, member C2; aldo-keto reductase family 1, member C21; aldo-keto reductase family 1, member C6; BABP; C9; chlordecone reductase homolog HAKRC; Chlordecone reductase homolog HAKRD; DD; DD/BABP; DD1; DD1/DD2; DD2; DD-2; DDH; DDH1; DDH2; dihydrodiol dehydrogenase 1; dihydrodiol dehydrogenase 1/2; dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase; Dihydrodiol dehydrogenase 2; dihydrodiol dehydrogenase 2; bile acid binding protein; 3-alpha hydroxysteroid dehydrogenase, type III; dihydrodiol dehydrogenase isoform DD1; Dihydrodiol dehydrogenase/bile acid-binding protein; H-37; HAKRC; HAKRD; HBAB; hepatic dihydrodiol dehydrogenase; high-affinity hepatic bile acid-binding protein; Hsd17b5; Indanol dehydrogenase; MBAB; MCDR2; pseudo-chlordecone reductase; SRXY8; TDD; testicular 17,20-desmolase deficiency; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type 5 17beta-hydroxysteroid dehydrogenase; type II 3-alpha-hydroxysteroid dehydrogenase; type II dihydrodiol dehydrogenase; Type III 3-alpha-hydroxysteroid dehydrogenase
Gene Symbols AKR1C1, Akr1c2
Host Species Rabbit
Immunogen E. coli-derived human AKR1C1/C2 recombinant protein (Position: M1-K123).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1645, 1646, 291283, 307092
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.