Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKR7A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100382
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100382 Supplier Invitrogen™ Supplier No. PA5100382
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AKR7A3.

Aldo-keto reductases, such as AKR7A3, are involved in the detoxification of aldehydes and ketones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR7A3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AKR7A3
Gene Accession No. O95154, P38918
Gene Alias Afar; AFAR2; AFB1 aldehyde reductase 2; AFB1-AR; AFB1-AR 2; aflatoxin aldehyde reductase; aflatoxin B1 aldehyde reductase 2; aflatoxin B1 aldehyde reductase member 1; Aflatoxin B1 aldehyde reductase member 3; Akr7a1; AKR7A3; aldo-keto reductase family 7 member A3; aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase); rAFAR1
Gene Symbols AKR7A3
Host Species Rabbit
Immunogen A synthesized peptide derived from human AKR7A3(Accession O95154), corresponding to amino acid residues T206-Y256.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22977, 26760
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.