Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Multispecies AKT1 (Total) InstantOne™ ELISA Kit

Catalog No. 501128670
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-8670 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-8670 Supplier Invitrogen™ Supplier No. 858604711
Add to Cart
Edge
Add to Cart

ELISA

The InstantOne™ ELISA is specifically engineered for accurate measurement of total human, mouse, and hamster AKT 1 in cell lysates. Detection of rat AKT 1 is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

AKT1 (PKB alpha) is a serine/threonine kinase that regulates cell survival. The activated enzyme inhibits apoptosis and stimulates cell cycle progression by phosphorylating numerous targets in various cell types, including cancer cells. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P31749
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Target Species Human, Rat, Mouse
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 207
Gene Symbol AKT,CWS6,PKB,PKB-ALPHA,PRKBA,RAC,RAC-ALPHA
Kit Contents Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 20 μL
Storage Requirements 2°C to 8°C
Synonym Protein kinase B, PKB, RAC-alpha serine/threonine-protein kinase
Target AKT1
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 10 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.