Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT Inhibitor VIII 97.0+%, TCI America™
SDP

Catalog No. A3153100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3153100MG 100 mg
A315325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3153100MG Supplier TCI America Supplier No. A3153100MG

Chemical Identifiers

CAS 612847-09-3
Molecular Formula C34H29N7O
Molecular Weight (g/mol) 551.654
MDL Number MFCD09026909
InChI Key BIWGYFZAEWGBAL-UHFFFAOYSA-N
Synonym 1,3-Dihydro-1-[1-[4-(6-phenyl-1H-imidazo[4,5-g]quinoxalin-7-yl)phenylmethyl]-4-piperidinyl]-2H-benzimidazol-2-one, Akti-1/2
PubChem CID 10196499
IUPAC Name 3-[1-[[4-(7-phenyl-3H-imidazo[4,5-g]quinoxalin-6-yl)phenyl]methyl]piperidin-4-yl]-1H-benzimidazol-2-one
SMILES C1CN(CCC1N2C3=CC=CC=C3NC2=O)CC4=CC=C(C=C4)C5=NC6=CC7=C(C=C6N=C5C8=CC=CC=C8)N=CN7

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 551.65
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material AKT Inhibitor VIII
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.