Learn More
CPC Scientific H-Ala-Arg-Lys-Arg-Glu-Arg-Thr-Tyr-Ser-Phe-Gly-His-His-Ala-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific HIST079B
Encompass
The native peptide ARKRERTYSFGHHA is a synthetic substrate for AKT/PKB/Rac-protein kinases. Phophorylation is at the Ser site (Km = 3.9 ?M). It also competitively inhibits histone H2B phosphorylation (Ki = 12 ?M) by AKT.ONE-LETTER SEQUENCE: ARKRERTYSFGHHAMOLECULAR FORMULA: C74H114N28O20MOLECULAR WEIGHT:1715.87STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [324029-01-8], RESEARCH AREA: MiscellaneousREFERENCES: Obata, T. et al. J. Biol. Chem. 275, 36108 (2000) Chan, TO. et al. Ann. Rev. Biochem. 68, 965 (1999)
Catalog No. 50-210-9522
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.