Learn More
Invitrogen™ AKT3 Monoclonal Antibody (66C1247.1)

Description
The sequence is identical in human, mouse, rat, cow, dog, and chicken. Suggested positive control: human kidney protein, antigen standard for AKT3 (transient overexpression lysate). Immunogen sequence: CSPTSQIDNI GEEEMDAS.
Specifications
Specifications
| Antigen | AKT3 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 66C1247.1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | AKT3 |
| Gene Accession No. | Q63484, Q9WUA6, Q9Y243 |
| Gene Alias | AI851531; AKT serine/threonine kinase 3; Akt3; AKT3 kinase; akt3a; D930002M15Rik; DKFZP434N0250; MPPH; MPPH2; Nmf350; PKB gamma; PKBG; PKB-GAMMA; PRKBG; Protein kinase Akt-3; protein kinase B gamma; protein kinase B, gamma; RAC-gamma; RAC-gamma serine/threonine protein kinase; RAC-gamma serine/threonine-protein kinase; RAC-PK-gamma; si:ch211-221n23.2; STK-2; thymoma viral proto-oncogene 3; v-akt murine thymoma viral oncogene 3; v-akt murine thymoma viral oncogene homolog 3; v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma); v-akt murine thymoma viral oncogene homolog 3a; v-akt murine thymoma viral oncogene-like 3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.