Learn More
AktS1/PRAS40 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301202AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
The Akt signaling pathway contributes to the regulation of apoptosis after a variety of cell death signals. AKT1S1, also known as PRAS40, is a proline-rich substrate of the kinase AKT1 and is thought to play a role in neuroprotection mediated by nerve growth factor (NGF) after transient focal cerebral ischemia. AKT1S1 is also a substrate and potential regulator of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Treatment with the insulin-like growth factor-1 (IGF1) can indusce the phosphorylation of AKT1S1 via the PI3K/AKT signaling pathway in PC12 cells.Specifications
| AktS1/PRAS40 | |
| Polyclonal | |
| Unconjugated | |
| AKT1S1 | |
| 40 kDa proline-rich AKT substrate; OTTHUMP00000196756; OTTHUMP00000196760; proline-rich AKT1 substrate 1; proline-rich Akt substrate, 40 kDa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84335 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q96B36 | |
| AKT1S1 | |
| Between 225 and 256 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.