Learn More
Advanced Biomatrix Akura 96-Well Spheroid and Organoid Microplate, 5 pack
Supplier:  Advanced Biomatrix 54375EA
Encompass_Preferred
The Akura 96-well Spheroid and Organoid Microplate is designed for lossless spheroid/organoid formation and handling as well as reliable culturing and analysis. The package includes 5 individually wrapped Akura 96 Spheroid Microplates, made of Cyclic olefin polymer, high-efficiency ULA coating, sterile, and includes a lid. Imaging speed and resolution are enhanced through defined 1 mm quick-reference region of interest and a flat bottom well to minimize optical aberration and ensure constant spheroid z-position. It features a unique SureXchange ledge engineered to prevent unintended spheroid/organoid aspiration while leaving a minimal residual volume (5-7 µl) for >90% medium removal using manual and automated pipetting. ANSI/SLAS standard format for full compatibility with automated liquid handling systems.
Catalog No. NC2410853
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.