Learn More
Thermo Scientific™ Al-23 Tube, One End Closed
Specifications
Specifications
| Quantity | 400 mm |
| Diameter (Metric) Inside | 6 mm |
| Diameter (Metric) Outside | 10 mm |
| Form | Solid |
Frequently Asked Questions (FAQs)
The maximum working temperature for the Al-23 Tube is 1950 degrees C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.