Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AL 8697

Catalog No. 475350
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-535-0 50 mg
47-531-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-535-0 Supplier Tocris Bioscience™ Supplier No. 4753/50
This item is not returnable. View return policy

Potent and selective p38α inhibitor

AL 8697 is a potent and selective p38α inhibitor (IC50 = 6 nM). Exhibits 14-fold greater inhibition of p38α compared to p38β; also displays 300-fold selectivity for p38α compared to a panel of 91 kinases. Exhibits anti-inflammatory properties.

Specifications

Purity >98%
Quantity 50 mg
Product Type AL 8697
Molecular Weight (g/mol) 402.41
Inhibitors AL 8697

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.