Learn More
Carbosynth LLC. Ala-Ala-Phe-CH2Cl [AAF-CMK] (5 mg vial), 1ea, MW: 339.82, Molecular formula: C16H22N3O3Cl, CAS# 102129-66-8
SDP
Supplier:  Carbosynth LLC. IAA3202V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: (L-Alanyl-L-Alanyl-L-Phenylalanyl)Chloromethane, Application: Inhibitor for Tripeptidyl Peptidase I and II (Component of Giant Protease with Some Proteasome Function), Chymotrypsin, and Chymase; TPP1 and TPPII ibhibitor, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20C, References: R. Glas, M. Bogyo, J.S. McMaster, M. Gaczynska, and H.L. Ploegh, Nature, 392, 618 (1998)., E. Geier, G. Pfeifer, M. Wilm, M. Lucchiari-Hartz, W. Baurmeister, K. Eichmann, and G. Niedermann, Science, 283, 978 (1999)., L.A. Johnson, K.E. Moon, and M. Eisenberg, Biochem. Biophys. Acta, 953, 269 (1988)
Catalog No. 50-168-5801
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.