Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alachlor Solution, 5000μg/mL in Methanol, Ultra Scientific

Catalog No. usepa1068
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1068 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1068 Supplier Agilent Technologies Supplier No. EPA10681
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15972-60-8 , 67-56-1
Molecular Formula C14H20ClNO2
Molecular Weight (g/mol) 269.769
InChI Key XCSGPAVHZFQHGE-UHFFFAOYSA-N
PubChem CID 2078
ChEBI CHEBI:2533
IUPAC Name 2-chloro-N-(2,6-diethylphenyl)-N-(methoxymethyl)acetamide
SMILES CCC1=C(C(=CC=C1)CC)N(COC)C(=O)CCl

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material Alachlor Solution, 5000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.