Learn More
Description
Peptide sequence: EHNNELVTGS SYESPPPDFR GQWINLPVLQ LTKDPLKTPG RLDHGTRTAF.
Specifications
Specifications
| Antigen | Aladin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | AAAS |
| Gene Accession No. | Q9NRG9 |
| Gene Alias | AAA; Aaas; AAASb; achalasia, adrenocortical insufficiency, alacrimia; achalasia, adrenocortical insufficiency, alacrimia (Allgrove, triple-A); ADRACALA; Adracalin; ALADIN; aladin WD repeat nucleoporin; Allgrove, triple-A; D030041N15Rik; GL003 |
| Gene Symbols | AAAS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
